Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153056.2 | NP_694696.2 | 10 | 1231 | ||
GenBank mRNA | AK135484.1 | - | 8 | 1443 | ||
AY251540.1 | - | 10 | 1207 | |||
BC026403.1 | - | 10 | 1196 | 4209498 | ||
BC026637.1 | - | 9 | 1272 | |||
BC026650.1 | - | 10 | 1199 | 4987746 | ||
BC040759.1 | - | 10 | 1196 | 4209498 | ||
BC080304.1 | - | 7 | 2791 |