Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC024730.1 | - | 8 | 1070 | ||
BC024981.1 | - | 8 | 1070 | |||
BC033398.1 | - | 8 | 1070 |