Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tom1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039092.3 | NP_001034181.1 | 2 | 189 | ||
NM_001039093.1 | NP_001034182.1 | 2 | 189 | |||
NM_153080.2 | NP_694720.2 | 2 | 189 | |||
GenBank mRNA | AF467887.1 | - | 2 | 169 | ||
AK036788.1 | - | 2 | 165 | |||
AK043095.1 | - | 2 | 169 | |||
AK054531.1 | - | 2 | 157 | |||
AK166040.1 | - | 2 | 189 | |||
BC062947.1 | - | 2 | 153 | 30536491 | ||
BC127266.1 | - | 2 | 155 | 30357295 |