Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cadm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153112.3 | NP_694752.1 | 2 | 159 | ||
GenBank mRNA | AK143204.1 | - | 2 | 160 | ||
AY059394.1 | - | 2 | 83 |