Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153166.2 | NP_694806.1 | 1 | 163 | ||
GenBank mRNA | AK044042.1 | - | 1 | 89 | ||
AK147529.1 | - | 1 | 86 | |||
BC036971.1 | - | 1 | 174 | 4527212 |