Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153168.2 | NP_694808.1 | 2 | 294 | ||
GenBank mRNA | AJ428066.1 | - | 2 | 294 | ||
AK032695.1 | - | 2 | 299 | |||
AK035538.1 | - | 2 | 289 | |||
AK037012.1 | - | 2 | 301 | |||
AK045640.1 | - | 2 | 253 | |||
AK053667.1 | - | 2 | 288 | |||
BC053916.1 | - | 2 | 280 | |||
BC080303.1 | - | 2 | 181 | 5695617 | ||
BC111892.1 | - | 2 | 265 | 40060998 | ||
BC112412.1 | - | 2 | 263 | 40060997 | ||
BC138206.1 | - | 2 | 268 | 8861225 | ||
BC145982.1 | - | 2 | 268 | 40131285 |