Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc36a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153170.3 | NP_694810.2 | 5, 6 | 636 | ||
GenBank mRNA | AF453744.1 | - | 5, 6 | 589 | ||
AF512429.1 | - | 5, 6 | 659 | |||
AK041041.1 | - | 5, 6 | 628 | |||
AK079053.1 | - | 5, 6 | 601 | |||
AK086373.1 | - | 5, 6 | 637 | |||
AK156409.1 | - | 5, 6 | 637 | |||
AK160394.1 | - | 5, 6 | 601 | |||
AY228366.1 | - | 5, 6 | 586 | |||
BC044800.1 | - | 5, 6 | 636 | 4189221 |