Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spg7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153176.4 | NP_694816.3 | 2 | 247 | ||
GenBank mRNA | AF512565.1 | - | 2 | 279 | ||
AF547215.1 | - | 2 | 576 | |||
AK145540.1 | - | 2 | 248 | |||
BC051051.1 | - | 2 | 181 | |||
BC055488.1 | - | 92 | ||||
BC138141.1 | - | 2 | 474 | 8861160 |