Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ago4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153177.3 | NP_694817.2 | 2 | 371 | ||
GenBank mRNA | AB081474.1 | - | 2 | 116 | ||
AK030018.1 | - | 2 | 387 | |||
BB616446.1 | - | 2 | 371 | |||
BC096023.1 | - | 2 | 153 | 30607807 |