Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153179.3 | NP_694819.2 | 4, 5 | 512 | ||
GenBank mRNA | AK052698.1 | - | 4, 5 | 521 | ||
AK143992.1 | - | 4, 5 | 496 | |||
AY130764.1 | - | 4, 5 | 504 | |||
AY438374.1 | - | 4, 5 | 386 | |||
BB661186.1 | - | 4, 5 | 521 |