Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctdsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153088.2 | NP_694728.1 | 3 | 555 | ||
GenBank mRNA | AK089478.1 | - | 3 | 463 | ||
AK167390.1 | - | 3 | 313 | |||
AY028804.1 | - | 3 | 580 | |||
BC049184.1 | - | 3 | 415 | 6439402 | ||
BC058085.1 | - | 2 | 742 | |||
BC065158.1 | - | 3 | 290 | 5717102 | ||
BC079638.1 | - | 3 | 290 | 5717102 |