Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phospho1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153104.3 | NP_694744.1 | 3 | 634 | ||
GenBank mRNA | AJ457190.1 | - | 3 | 647 |