Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153114.2 | NP_694754.1 | 5 | 591 | ||
GenBank mRNA | AK083090.1 | - | 4 | 2879 | ||
AY078071.1 | - | 4 | 339 | |||
BC100323.1 | - | 5 | 591 | 30788363 | ||
BC132494.1 | - | 4 | 510 | 40130771 | ||
BC132498.1 | - | 4 | 510 | 40130775 |