Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153126.2 | NP_694766.1 | 2, 3 | 322 | ||
GenBank mRNA | AK036294.1 | - | 2, 3 | 270 | ||
AK040070.1 | - | 2, 3 | 277 | |||
AK054029.1 | - | 2, 3 | 343 | |||
AK087445.1 | - | 2, 3 | 280 | |||
AK142682.1 | - | 2, 3 | 334 | |||
AK149825.1 | - | 2, 3 | 149 | |||
BC034516.1 | - | 2, 3 | 322 | 4458024 |