Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irgq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153134.3 | NP_694774.3 | 3 | 1281 | ||
GenBank mRNA | AF322649.1 | - | 2 | 1070 | ||
AK051117.1 | - | 3 | 1191 | |||
AK210871.1 | - | 1 | 210 | |||
BC056347.1 | - | 3 | 1240 | 5705226 | ||
BC080294.1 | - | 3 | 1219 | 6856174 |