Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35e4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153142.3 | NP_694782.1 | 2 | 2446 | ||
GenBank mRNA | AK039786.1 | - | 2 | 2447 | ||
AK218784.1 | - | 1 | 72 | |||
BC020181.1 | - | 2 | 1480 | 4488513 | ||
BC095957.1 | - | 2 | 354 |