Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003.2 | NP_000994.1 | 4 | 443 | ||
NM_213725.1 | NP_998890.1 | 3 | 368 | |||
GenBank mRNA | AK026579.1 | - | 4 | 443 | ||
AK130958.1 | - | 4 | 446 | |||
AY303789.1 | - | 4 | 444 | |||
BC003369.1 | - | 4 | 426 | 2900846 | ||
BC007590.1 | - | 4 | 414 | 3343021 | ||
CR450339.1 | - | 4 | 314 | |||
CR542209.1 | - | 4 | 314 | |||
M17886.1 | - | 4 | 443 |
Gene Symbol: TTC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017024801.1 | XP_016880290.1 | 10 | 1803 | ||
XM_017024802.1 | XP_016880291.1 | 10 | 1861 |