Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stoml3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153156.1 | NP_694796.1 | 3, 4 | 218 | ||
GenBank mRNA | AB085692.1 | - | 3, 4 | 218 | ||
BC058224.1 | - | 264 | ||||
BC080859.1 | - | 247 | ||||
BC138667.1 | - | 3, 4 | 299 | 8861688 | ||
BC138668.1 | - | 3, 4 | 299 | 8861689 |