Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153159.2 | NP_694799.1 | 2 | 310 | ||
GenBank mRNA | AK142501.1 | - | 2 | 302 | ||
AK149698.1 | - | 2 | 309 | |||
AK152196.1 | - | 2 | 267 | |||
AK154363.1 | - | 2 | 284 | |||
AK161150.1 | - | 2 | 266 | |||
AK161733.1 | - | 2 | 265 | |||
AK163109.1 | - | 2 | 299 | |||
AK172357.1 | - | 2 | 267 | |||
AY920404.1 | - | 1 | 161 | |||
BC036563.1 | - | 2 | 269 | 5254681 |