Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039059.1 | NP_001034148.1 | 3 | 492 | ||
NM_001039060.1 | NP_001034149.1 | 4 | 543 | |||
NM_001039061.1 | NP_001034150.1 | 2 | 587 | |||
NM_153165.2 | NP_694805.1 | 4 | 543 | |||
GenBank mRNA | AK078112.1 | - | 2 | 528 | ||
AK081546.1 | - | 3 | 496 | |||
AK133055.1 | - | 3 | 257 | |||
AK169500.1 | - | 4 | 486 | |||
BC036978.1 | - | 4 | 480 | |||
BC141165.1 | - | 3 | 257 | 9055777 |