Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hus1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153072.2 | NP_694712.1 | 1 | 508 | ||
GenBank mRNA | AF508546.1 | - | 1 | 421 | ||
AK133107.1 | - | 1 | 510 | |||
BC125607.1 | - | 1 | 501 | 40130122 | ||
BC125609.1 | - | 1 | 501 | 40130124 |