Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ehbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252515.1 | NP_001239444.1 | 10, 11 | 1494 | ||
NM_153078.4 | NP_694718.4 | 10, 11 | 1419 | |||
GenBank mRNA | AF424697.1 | - | 9, 10 | 1269 | ||
AK076625.1 | - | 1246 | ||||
AK173055.1 | - | 10, 11 | 1495 |