Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153103.2 | NP_694743.2 | 14, 15 | 1713 | ||
GenBank mRNA | AB074017.1 | - | 12, 13 | 1343 | ||
AK046574.1 | - | 9, 10 | 954 | |||
AK173007.1 | - | 13, 14 | 1420 | |||
BC029286.1 | - | 14, 15 | 1700 |