Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgif2lx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153109.1 | NP_694749.1 | 1 | 833 | ||
GenBank mRNA | AF434662.1 | - | 1 | 833 | ||
AK132935.1 | - | 1 | 842 |