Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153131.3 | NP_694771.1 | 10 | 1823 | ||
GenBank mRNA | AJ487852.1 | - | 10 | 1824 | ||
BC058084.1 | - | 9 | 1676 | 6813463 |