Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286750.1 | NP_001273679.1 | 6 | 2985 | ||
NM_153157.3 | NP_694797.1 | 6 | 3136 | |||
NM_153458.3 | NP_703188.1 | 6 | 3389 | |||
GenBank mRNA | AF442824.1 | - | 6 | 3137 | ||
AF442825.1 | - | 6 | 3105 | |||
AK038569.1 | - | 6 | 3336 | |||
AK044325.1 | - | 6 | 3192 | |||
AK045482.1 | - | 6 | 3338 | |||
AK049612.1 | - | 6 | 3390 | |||
AK137414.1 | - | 6 | 3191 |