Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153179.3 | NP_694819.2 | 66 | 11919 | ||
GenBank mRNA | AY130764.1 | - | 66 | 11911 | ||
AY438374.1 | - | 66 | 11796 |