Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd3b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153193.3 | NP_694873.2 | 6 | 914 | ||
GenBank mRNA | BC026757.1 | - | 6 | 914 | 5136428 | |
BC040397.1 | - | 957 | 4222081 | |||
M75886.1 | - | 1 | 318 |