Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp10d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153389.3 | NP_700438.3 | 21 | 3914 | ||
GenBank mRNA | AJ441079.1 | - | 21 | 3914 | ||
AK036646.1 | - | 20 | 3576 | |||
AK133052.1 | - | 10 | 1365 | |||
AK144558.1 | - | 3 | 299 | |||
BC029551.1 | - | 4 | 373 | |||
BC064739.1 | - | 5 | 466 | |||
BC141328.1 | - | 21 | 3765 | 9055940 |