Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023.3 | NP_001014.1 | 4 | 423 | ||
NM_001146227.1 | NP_001139699.1 | 4 | 423 | |||
GenBank mRNA | AK130961.1 | - | 4 | 348 | ||
AK301342.1 | - | 4 | 352 | |||
AK311808.1 | - | 4 | 348 | |||
AV716603.1 | - | 352 | ||||
BC007507.2 | - | 4 | 332 | 3029762 | ||
BC087850.1 | - | 4 | 318 | 30386230 | ||
BQ064297.1 | - | 4 | 333 | |||
CB138358.1 | - | 4 | 423 | |||
L06498.1 | - | 4 | 338 |
Gene Symbol: RPS20P27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC071734.1 | - | 1 | 638 |