Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153394.2 | NP_700443.2 | 3, 4 | 708 | ||
GenBank mRNA | AF488740.1 | - | 2, 3 | 232 | ||
AK141990.1 | - | 3, 4 | 753 | |||
BC059227.1 | - | 3, 4 | 708 | 6400663 |