Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mical3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270475.1 | NP_001257404.1 | 2 | 468 | ||
NM_153396.3 | NP_700445.2 | 2 | 434 | |||
GenBank mRNA | AF536756.1 | - | 2 | 397 | ||
AK048201.1 | - | 2 | 446 | |||
AK122497.2 | - | 2 | 293 | |||
AK132669.1 | - | 2 | 418 | |||
BC138257.1 | - | 2 | 395 | 8861276 | ||
BC145214.1 | - | 2 | 395 | |||
CJ105256.1 | - | 2 | 469 |