Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253731.1 | NP_001240660.1 | 3 | 497 | ||
NM_153414.4 | NP_700463.2 | 3 | 465 | |||
GenBank mRNA | AK035769.1 | - | 3 | 209 | ||
AK038979.1 | - | 3 | 198 | |||
AK077634.1 | - | 3 | 466 | |||
AK136992.1 | - | 3 | 192 | |||
AK150195.1 | - | 3 | 192 | |||
BC028953.1 | - | 2 | 155 | 5362016 | ||
BC055700.1 | - | 3 | 164 | 6808816 | ||
BY195822.1 | - | 3 | 230 |