Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ammecr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242430.1 | NP_001229359.1 | 3 | 868 | ||
NM_153515.4 | NP_705735.1 | 3 | 667 | |||
GenBank mRNA | AK030009.1 | - | 3 | 875 | ||
AK054131.1 | - | 3 | 666 | |||
AK133018.1 | - | 3 | 869 | |||
AK151060.1 | - | 3 | 668 | |||
AK151257.1 | - | 3 | 568 | |||
AK192354.1 | - | 1 | 136 | |||
AK196122.1 | - | 1 | 135 | |||
BC012310.1 | - | 2 | 272 | |||
BC034661.1 | - | 3 | 408 | 5137474 | ||
BC056948.1 | - | 3 | 611 | 6827318 |