Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dis3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172157.1 | NP_001165628.1 | 3 | 269 | ||
NM_153530.2 | NP_705758.1 | 3 | 269 | |||
GenBank mRNA | AK029652.1 | - | 3 | 246 | ||
AK031180.1 | - | 3 | 254 | |||
AK040613.1 | - | 3 | 246 | |||
AK050762.1 | - | 3 | 270 | |||
AK079867.1 | - | 3 | 266 | |||
BB654969.1 | - | 3 | 270 | |||
BC036177.1 | - | 3 | 230 | 5004341 | ||
CJ063044.1 | - | 3 | 248 |