Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phldb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153537.4 | NP_705765.3 | 15, 16 | 3238 | ||
GenBank mRNA | AK052352.1 | - | 4, 5 | 598 | ||
AK122336.1 | - | 13, 14 | 3124 | |||
BC053931.1 | - | 10, 11 | 1367 | |||
BC058712.1 | - | 15, 16 | 3234 | 6842596 |