Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153542.1 | NP_705770.1 | 4 | 376 | ||
GenBank mRNA | AK136380.1 | - | 4 | 403 | ||
AK139384.1 | - | 4 | 409 | |||
AK145920.1 | - | 4 | 393 | |||
AK150142.1 | - | 4 | 377 | |||
AK150246.1 | - | 4 | 377 | |||
BC036304.1 | - | 4 | 376 | 3968534 |