Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153555.2 | NP_705783.1 | 4 | 329 | ||
GenBank mRNA | AK035701.1 | - | 4 | 228 | ||
AK047316.1 | - | 4 | 233 | |||
AK098114.1 | - | 4 | 348 | |||
AK146825.1 | - | 4 | 256 | |||
AK162622.1 | - | 4 | 351 | |||
AK164799.1 | - | 4 | 256 | |||
AK167059.1 | - | 4 | 205 | |||
AK199445.1 | - | 2 | 71 | |||
AK203373.1 | - | 3 | 71 | |||
BB851975.1 | - | 5 | 330 | |||
BC023704.1 | - | 5 | 270 | 5345701 | ||
BC023804.1 | - | 5 | 383 | 5352941 | ||
BC078641.1 | - | 4 | 1241 |