Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam102a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153560.4 | NP_705788.1 | 2 | 889 | ||
GenBank mRNA | AK049032.1 | - | 2 | 893 | ||
AK131145.1 | - | 2 | 916 | |||
AK157331.1 | - | 2 | 519 | |||
AK157490.1 | - | 3 | 311 | |||
AK158435.1 | - | 2 | 516 | |||
AK165751.1 | - | 2 | 519 | |||
AK170536.1 | - | 2 | 917 | |||
AK170847.1 | - | 2 | 510 | |||
AK170951.1 | - | 2 | 516 | |||
AK172536.1 | - | 2 | 516 | |||
AK205749.1 | - | 2 | 42 | |||
BC023752.1 | - | 2 | 334 | 5354455 | ||
BC031157.1 | - | 2 | 152 | 4976773 |