Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164833.1 | NP_001158305.1 | 3 | 429 | ||
NM_001164834.1 | NP_001158306.1 | 3 | 429 | |||
NM_153400.4 | NP_700449.2 | 3 | 429 | |||
GenBank mRNA | AB094663.1 | - | 3 | 349 | ||
AB094664.1 | - | 3 | 349 | |||
AK141196.1 | - | 3 | 388 | |||
AK158334.1 | - | 3 | 407 | |||
AY044240.2 | - | 3 | 605 | |||
BC106963.1 | - | 3 | 236 | |||
BC106964.1 | - | 3 | 236 | 40039097 |