Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Specc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145826.1 | NP_001139298.1 | 7 | 1744 | ||
NM_153406.3 | NP_700455.3 | 4 | 1515 | |||
GenBank mRNA | AB093236.1 | - | 7 | 1744 | ||
AK035500.1 | - | 4 | 1517 | |||
AK052716.1 | - | 4 | 1516 | |||
AY884297.1 | - | 4 | 1516 | |||
BC037464.1 | - | 1 | 602 | 4194868 | ||
BC072595.1 | - | 3 | 1354 | 6408832 |