Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199146.1 | NP_001186075.1 | 2 | 308 | ||
NM_001199147.1 | NP_001186076.1 | 2 | 377 | |||
NM_153410.5 | NP_700459.2 | 2 | 308 | |||
GenBank mRNA | AK048691.1 | - | 2 | 302 | ||
AK082985.1 | - | 2 | 251 | |||
AK084631.1 | - | 2 | 250 | |||
AK209240.1 | - | 1 | 67 | |||
BC026486.1 | - | 2 | 368 | |||
BC071197.1 | - | 2 | 374 | 6465025 | ||
CA317518.1 | - | 2 | 326 | |||
CK618701.1 | - | 2 | 377 |