Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153526.5 | NP_705746.1 | 4 | 746 | ||
GenBank mRNA | AF527630.1 | - | 4 | 760 | ||
AK088380.1 | - | 4 | 793 | |||
AK135941.1 | - | 4 | 797 | |||
AK135989.1 | - | 4 | 799 | |||
AK139913.1 | - | 4 | 796 | |||
AK151186.1 | - | 4 | 796 | |||
BC079559.1 | - | 4 | 746 | |||
BC132167.1 | - | 4 | 754 | 40130444 | ||
BC132499.1 | - | 4 | 754 | 40130776 |