Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001063.3 | NP_001054.1 | 3, 4 | 640 | ||
XM_017007089.1 | XP_016862578.1 | 9, 10 | 1990 | |||
XM_017007090.1 | XP_016862579.1 | 9, 10 | 2983 | |||
GenBank mRNA | AK222755.1 | - | 3, 4 | 380 | ||
AK293842.1 | - | 3, 4 | 380 | |||
AK295334.1 | - | 3, 4 | 380 | |||
AK295419.1 | - | 2, 3 | 207 | |||
AK303753.1 | - | 3, 4 | 380 | |||
BC059367.1 | - | 3, 4 | 378 | 5264299 | ||
BX648533.1 | - | 3, 4 | 157 | |||
CR936810.1 | - | 8, 9 | 2061 | |||
DQ923758.1 | - | 3, 4 | 371 | |||
GQ472199.1 | - | 3, 4 | 378 | |||
M12530.1 | - | 3, 4 | 362 | |||
S95936.1 | - | 3, 4 | 411 |