Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153534.2 | NP_705762.2 | 2, 3 | 674 | ||
GenBank mRNA | AK122432.1 | - | 2, 3 | 489 | ||
AK132312.1 | - | 2, 3 | 675 | |||
BC037107.1 | - | 2, 3 | 431 | 5367175 |