Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TKT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001064.3 | NP_001055.1 | 4, 5 | 612 | ||
NM_001135055.2 | NP_001128527.1 | 4, 5 | 612 | |||
GenBank mRNA | AK093667.1 | - | 3, 4 | 271 | ||
AK223614.1 | - | 4, 5 | 520 | |||
AK289454.1 | - | 4, 5 | 488 | |||
AK303191.1 | - | 5, 6 | 479 | |||
BC002433.2 | - | 4, 5 | 433 | 3346980 | ||
BC009970.2 | - | 4, 5 | 510 | 4310396 | ||
BC024026.2 | - | 2, 3 | 181 | |||
BX647248.1 | - | 4, 5 | 498 | |||
EU794667.1 | - | 4, 5 | 510 | |||
KU178361.1 | - | 4, 5 | 440 | |||
KU178362.1 | - | 4, 5 | 440 | |||
L12711.1 | - | 4, 5 | 538 | |||
U55017.1 | - | 4, 5 | 440 | |||
X67688.1 | - | 5, 6 | 510 |