Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153391.2 | NP_700440.2 | 16 | 1807 | ||
GenBank mRNA | AK052303.1 | - | 16 | 1787 | ||
AK129413.1 | - | 8 | 896 | |||
AK147970.1 | - | 16 | 1807 | |||
AK166172.1 | - | 16 | 1773 | |||
AY029258.1 | - | 16 | 1877 | |||
BC060134.1 | - | 16 | 1801 | |||
BC082301.1 | - | 15 | 1687 |