Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153405.2 | NP_700454.1 | 8 | 1331 | ||
GenBank mRNA | AB036992.1 | - | 8 | 1247 | ||
AK089975.1 | - | 8 | 1224 | |||
AK090084.1 | - | 8 | 1300 | |||
AK140861.1 | - | 8 | 1332 | |||
AY152391.1 | - | 8 | 1331 | |||
BC057890.1 | - | 8 | 1242 | 4979982 | ||
CB597326.1 | - | 2 | 176 |