Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153501.2 | NP_705721.2 | 2 | 534 | ||
GenBank mRNA | AK016755.1 | - | 4 | 557 | ||
AK082821.1 | - | 2 | 540 | |||
AK154080.1 | - | 2 | 498 | |||
AK159175.1 | - | 2 | 525 | |||
AK167861.1 | - | 2 | 242 | |||
AK180859.1 | - | 1 | 52 | |||
AK186345.1 | - | 1 | 52 | |||
AK208629.1 | - | 1 | 52 | |||
AK211080.1 | - | 1 | 52 | |||
BC106184.1 | - | 2 | 488 | 30241166 |