Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153527.2 | NP_705755.2 | 4, 5 | 554 | ||
GenBank mRNA | AF419292.2 | - | 4, 5 | 699 | ||
AK005984.1 | - | 4, 5 | 571 | |||
AK139680.1 | - | 527 | ||||
BC048501.1 | - | 4, 5 | 554 | 6705915 |